This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04418286_s1
See other MIR600 GE Assays ›
Gene Symbol:
MIR600 STRBP
Gene Name:
microRNA mir-600
spermatid perinuclear RNA binding protein
Gene Aliases:
mml-mir-600
-
Chromosome Location:
Chr.15: 17564165 - 17564258 on Build Mmul_10
Chromosome Location:
Chr.15: 17415403 - 17566299 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh04418286_s1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIR600
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_032639.1 49 77
Gene Symbol: STRBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028834222.1 XP_028690055.1 13856 77
XM_028834223.1 XP_028690056.1 13968 77
XM_028834225.1 XP_028690058.1 12964 77
XM_028834227.1 XP_028690060.1 13556 77

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0