This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04382860_m1
See other EIF4EBP3 GE Assays ›
Gene Symbol:
EIF4EBP3 LOC698045
Gene Name:
eukaryotic translation initiation factor 4E binding protein 3
ankyrin repeat and KH domain-containing protein 1
Gene Aliases:
-
ANKHD1
Chromosome Location:
Chr.6: 137941586 - 137943485 on Build Mmul_10
Chromosome Location:
Chr.6: 137802853 - 137935653 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh04382860_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4EBP3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204096.2 NP_001191025.1 351 75
GenBank mRNA JV581847.1 - 204 75
Gene Symbol: LOC698045
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA JU475591.1 - 7962 75
JV636154.1 - 7911 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0