This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02890394_m1
See other STING1 GE Assays ›
Gene Symbol:
STING1
Gene Name:
stimulator of interferon response cGAMP interactor 1
Gene Aliases:
STING, TMEM173
Chromosome Location:
Chr.6: 136885904 - 136893781 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02890394_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STING1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015141010.2 XP_014996496.2 553 112
XM_028850089.1 XP_028705922.1 355 112
GenBank mRNA JU472368.1 - 525 112
MF622060.1 - 225 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6d4c5dbbff-7nlnn:80/100.66.128.142:80.
git-commit: 3992c1c7134a3a3ff95f78ba9fe94218727d3638
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0