This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02871011_m1
See other LOC711104 GE Assays ›
Gene Symbol:
LOC711104
Gene Name:
putative RNA-binding protein Luc7-like 2
Gene Aliases:
LUC7L2
Chromosome Location:
Chr.3: 165781692 - 165848890 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02871011_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC711104
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001108444.4 XP_001108444.1 1267 102
XM_015135024.2 XP_014990510.1 940 102
XM_015135025.2 XP_014990511.1 1272 102
XM_015135026.2 XP_014990512.1 3622 102
XM_015135028.2 XP_014990514.1 3142 102
XM_015135034.2 XP_014990520.1 1201 102
XM_015135036.2 XP_014990522.1 4726 102
XM_015135040.2 XP_014990526.1 1337 102
XM_028846365.1 XP_028702198.1 1956 102
GenBank mRNA JU319762.1 - 1154 102
JU471676.1 - 915 102
JV044892.1 - 1073 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0