This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02870918_m1
See other UBE2D4 GE Assays ›
Gene Symbol:
UBE2D4
Gene Name:
ubiquitin conjugating enzyme E2 D4 (putative)
Gene Aliases:
EGK_13741
Chromosome Location:
Chr.3: 71190537 - 71218771 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02870918_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2D4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002803298.3 XP_002803344.1 201 123
XM_015133660.2 XP_014989146.1 393 123
XM_015133661.2 XP_014989147.1 393 123
XM_015133662.2 XP_014989148.1 238 123
XM_015133663.2 XP_014989149.1 340 254
XM_015133667.2 XP_014989153.1 295 123
XM_028845846.1 XP_028701679.1 289 254
XM_028845849.1 XP_028701682.1 337 254
GenBank mRNA JU320239.1 - 184 123
JU475945.1 - 186 123
JV047336.1 - 161 123
JV471140.1 - 101 123

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015133663.2, XM_028845846.1, XM_028845849.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0