This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02869913_m1
See other MYRIP GE Assays ›
Gene Symbol:
MYRIP
Gene Name:
myosin VIIA and Rab interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.2: 95821979 - 96232760 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02869913_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYRIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001115677.4 XP_001115677.2 2713 67
XM_015131301.2 XP_014986787.2 2692 67
XM_015131302.2 XP_014986788.2 2632 67
XM_015131303.2 XP_014986789.2 2270 67
XM_028843982.1 XP_028699815.1 2518 67
GenBank mRNA JU323063.1 - 2499 67
JV527773.1 - 2438 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

myelin protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0