Assay Details
Gene Symbol: CA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099188.4 | XP_001099188.2 | 621 | 96 | ||
XM_015146262.2 | XP_015001748.1 | 621 | 96 | |||
XM_015146376.2 | XP_015001862.1 | 615 | 96 | |||
XM_028840191.1 | XP_028696024.1 | 621 | 96 | |||
GenBank mRNA | JU477123.1 | - | 519 | 96 | ||
JV048197.1 | - | 508 | 96 | |||
JV660825.1 | - | 526 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
