This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02804945_m1
See other BUD23 GE Assays ›
Gene Symbol:
BUD23
Gene Name:
BUD23 rRNA methyltransferase and ribosome maturation factor
Gene Aliases:
WBSCR22
Chromosome Location:
Chr.3: 46040468 - 46055343 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02804945_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BUD23
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266959.1 NP_001253888.1 295 96
XM_028845071.1 XP_028700904.1 421 96
GenBank mRNA JU332790.1 - 301 96
JU332791.1 - 301 96
JU332792.1 - 301 96
JU474210.1 - 301 96
JV043191.1 - 295 96
JV706194.1 - 317 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Macaca mulatta

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0