This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02795879_g1
See other RPS21 GE Assays ›
Gene Symbol:
RPS21
Gene Name:
ribosomal protein S21
Gene Aliases:
-
Chromosome Location:
Chr.10: 97543183 - 97544723 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02795879_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS21
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015148649.2 XP_015004135.1 139 67
XM_015148650.2 XP_015004136.2 139 67
XM_028828701.1 XP_028684534.1 153 67
GenBank mRNA JV527813.1 - 84 67

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ribosomal protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7887d5bf67-gxz2v:80/100.66.128.142:80.
git-commit: 1c5b77651f08bdd3ffd11596e5fe4ce144025943
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0