This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02787977_m1
See other ICAM2 GE Assays ›
Gene Symbol:
ICAM2
Gene Name:
intercellular adhesion molecule 2
Gene Aliases:
EGK_08876
Chromosome Location:
Chr.16: 60561289 - 60580883 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02787977_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ICAM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195389.1 NP_001182318.1 499 75
XM_015120046.2 XP_014975532.1 483 75
GenBank mRNA AF340054.1 - 282 75
CB551343.1 - 515 75
CN644791.1 - 141 75
JU496308.1 - 1523 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0