Assay Details
Gene Symbol: DDX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258162.1 | NP_001245091.1 | 664 | 64 | ||
XM_015120047.2 | XP_014975533.1 | 1009 | 64 | |||
XM_015120048.2 | XP_014975534.1 | 818 | 64 | |||
GenBank mRNA | JU333198.1 | - | 490 | 64 | ||
JU474511.1 | - | 664 | 64 | |||
JV046322.1 | - | 629 | 64 | |||
JV643960.1 | - | 676 | 64 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
