This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01062552_m1
See other CERT1 GE Assays ›
Gene Symbol:
CERT1
Gene Name:
ceramide transporter 1
Gene Aliases:
COL4A3BP
Chromosome Location:
Chr.6: 72244539 - 72393524 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01062552_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015140286.2 XP_014995772.1 1129 67
XM_015140287.1 XP_014995773.1 975 67
XM_015140288.2 XP_014995774.1 975 67
XM_028849517.1 XP_028705350.1 975 67
XM_028849519.1 XP_028705352.1 975 67
XM_028849520.1 XP_028705353.1 975 67
GenBank mRNA JU478502.1 - 811 67
JV704855.1 - 883 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transfer/carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0