This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01031101_m1
See other HDGFL2 GE Assays ›
Gene Symbol:
HDGFL2
Gene Name:
HDGF like 2
Gene Aliases:
HDGFRP2
Chromosome Location:
Chr.19: 4384234 - 4420446 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01031101_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDGFL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015122527.2 XP_014978013.1 1072 88
XM_028838604.1 XP_028694437.1 1054 88
XM_028838605.1 XP_028694438.1 1057 88
XM_028838606.1 XP_028694439.1 1037 88
XM_028838607.1 XP_028694440.1 1150 88
GenBank mRNA JU472454.1 - 938 88
JV043676.1 - 995 88
JV484698.1 - 586 88
JV635428.1 - 985 88
JV668638.1 - 996 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transcription cofactor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0