This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00979300_m1
See other MTFR1 GE Assays ›
Gene Symbol:
MTFR1
Gene Name:
mitochondrial fission regulator 1
Gene Aliases:
EGK_18996
Chromosome Location:
Chr.8: 66268475 - 66356763 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00979300_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTFR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015145458.2 XP_015000944.1 343 125
XM_015145459.2 XP_015000945.1 445 125
XM_015145460.2 XP_015000946.1 330 125
XM_015145461.2 XP_015000947.1 343 125
XM_015145462.2 XP_015000948.1 343 125
XM_015145465.2 XP_015000951.1 370 125
XM_028852701.1 XP_028708534.1 344 125
GenBank mRNA JU337258.1 - 302 125
JU470702.1 - 282 125
JV727580.1 - 284 125

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0