This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03630540_g1
See other LOC4339910 GE Assays ›
Gene Symbol:
LOC4339910
Gene Name:
ruBisCO large subunit-binding protein subunit beta, chloroplastic
Gene Aliases:
OSNPB_060114000
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03630540_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4339910
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015787279.2 XP_015642765.1 14 - 15 1886 70
XM_015787280.2 XP_015642766.1 14 - 15 1875 70
GenBank mRNA AK060117.1 - 8 - 9 875 70
AK062098.1 - 12 - 13 1549 70
AK101334.1 - 14 - 15 1881 70
EU325985.1 - 13 - 14 1792 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0