This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03627816_g1
See other LOC4336384 GE Assays ›
Gene Symbol:
LOC4336384 LOC4336150
Gene Name:
BRCA1-associated RING domain protein 1
2-oxoglutarate-dependent dioxygenase DAO-like
Gene Aliases:
OSNPB_040512400
DAO, OSNPB_040475600
Chromosome Location:
-
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
-
Assay ID Os03627816_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4336384
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015778572.2 XP_015634058.1 3 - 4 586 89
XM_015778573.2 XP_015634059.1 3 - 4 587 89
GenBank mRNA AK103203.1 - 3 - 4 583 89
Gene Symbol: LOC4336150
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK288722.1 - 2175 89

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0