This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03602018_m1
See other LOC4328383 GE Assays ›
Gene Symbol:
LOC4328383
Gene Name:
CBL-interacting protein kinase 26-like
Gene Aliases:
CIPK26, OJ9003_G05.12, OSNPB_020161000, OsCIPK26, OsJ_005311
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03602018_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4328383
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015769220.2 XP_015624706.1 1 - 2 470 106
GenBank mRNA AK111660.1 - 1 - 2 97 106

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015769220.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Oryza sativa Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0