Assay Details
Gene Symbol: LOC4326732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015766191.2 | XP_015621677.1 | 7 - 8 | 882 | 79 | |
XM_015766192.2 | XP_015621678.1 | 7 - 8 | 881 | 79 | ||
GenBank mRNA | AK064461.1 | - | 2 - 3 | 756 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
