Assay Details
Gene Symbol: LOC4337289 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780728.2 | XP_015636214.1 | 3 - 4 | 708 | 57 | |
GenBank mRNA | AK066532.1 | - | 3 - 4 | 714 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
