Assay Details
Gene Symbol: LOC4337211 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777875.2 | XP_015633361.1 | 12 - 13 | 1544 | 135 | |
XM_015777876.1 | XP_015633362.1 | 11 - 12 | 1496 | 135 | ||
GenBank mRNA | AK064227.1 | - | 11 - 12 | 1484 | 135 | |
AK071891.1 | - | 11 - 12 | 1504 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
