Assay Details
Gene Symbol: LOC4336481 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015780364.2 | XP_015635850.1 | 3 - 4 | 819 | 75 | |
GenBank mRNA | AK066705.1 | - | 3 - 4 | 790 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
