This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03535444_m1
See other LOC4333604 GE Assays ›
Gene Symbol:
LOC4333604
Gene Name:
chlorophyll(ide) b reductase NOL, chloroplastic-like
Gene Aliases:
NOL, NOL1, OSNPB_030654600, OsJ_11943, OsNOL
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03535444_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4333604
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015772787.2 XP_015628273.1 7 - 8 792 68
XM_015772788.2 XP_015628274.1 7 - 8 792 68
GenBank mRNA AB255026.1 - 7 - 8 670 68
AK287869.1 - 7 - 8 793 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

transfer/carrier protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0