Assay Details
Gene Symbol: LOC4328521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770868.2 | XP_015626354.1 | 8 - 9 | 1158 | 94 | |
GenBank mRNA | AK120718.1 | - | 7 - 8 | 1235 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
