Assay Details
Gene Symbol: LOC4328521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015770868.2 | XP_015626354.1 | 11 - 12 | 1416 | 86 | |
GenBank mRNA | AK120718.1 | - | 10 - 11 | 1493 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
