Assay Details
Gene Symbol: LOC4348056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015757484.2 | XP_015612970.1 | 5 - 6 | 1040 | 96 | |
GenBank mRNA | AK068217.1 | - | 5 - 6 | 929 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
