Assay Details
Gene Symbol: LOC4343652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792175.2 | XP_015647661.1 | 2 - 3 | 554 | 86 | |
GenBank mRNA | AK067599.1 | - | 2 - 3 | 400 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
