Assay Details
Gene Symbol: LOC4335601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015778796.2 | 2 - 3 | 282 | 142 | ||
XM_015778797.2 | 2 - 3 | 294 | 142 | |||
XM_015778798.2 | XP_015634284.1 | 2 - 3 | 281 | 142 | ||
XM_015778800.2 | XP_015634286.1 | 2 - 3 | 314 | 142 | ||
XM_015778801.2 | XP_015634287.1 | 2 - 3 | 207 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
