Assay Details
Gene Symbol: LOC4327645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015765933.2 | XP_015621419.1 | 4 - 5 | 617 | 99 | |
XM_015765935.2 | XP_015621421.1 | 4 - 5 | 617 | 99 | ||
XM_015765936.2 | XP_015621422.1 | 4 - 5 | 617 | 99 | ||
GenBank mRNA | AK100326.1 | - | 4 - 5 | 617 | 99 | |
GQ267545.1 | - | 3 - 4 | 402 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
