This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06769375_m1
See other LOC103345890 GE Assays ›
Gene Symbol:
LOC103345890
Gene Name:
histone-lysine N-methyltransferase SETD2-like
Gene Aliases:
-
Chromosome Location:
Chr.Unknown: 319286 - 331418 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06769375_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC103345890
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017339093.1 XP_017194582.1 811 83
XM_017339094.1 XP_017194583.1 793 83

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017339093.1, XM_017339094.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus

Panther Classification:

Molecular Function -

histone modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0