This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06764521_g1
See other LOC100356626 GE Assays ›
Gene Symbol:
LOC100356626
Gene Name:
cat eye syndrome critical region protein 2
Gene Aliases:
CECR2
Chromosome Location:
Chr.8: 38077560 - 38143847 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06764521_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100356626
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008259878.2 XP_008258100.1 1 - 2 198 70
XM_008259879.2 XP_008258101.1 1 - 2 198 70
XM_017343450.1 XP_017198939.1 1 - 2 199 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008259878.2, XM_008259879.2, XM_017343450.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0