This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc06672538_g1
See other LOC100340374 GE Assays ›
Gene Symbol:
LOC100340374
Gene Name:
sialic acid-binding Ig-like lectin 10
Gene Aliases:
SIGLEC10
Chromosome Location:
Chr.Unknown: 104709 - 110722 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc06672538_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100340374
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008252953.2 XP_008251175.1 1227 99
XM_008252954.2 XP_008251176.1 1131 99
XM_008252956.2 XP_008251178.1 1225 99

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Oryctolagus cuniculus

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9dd846b7f-s9zn4:80/100.66.128.142:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0