This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oc03823449_s1
See other BPGM GE Assays ›
Gene Symbol:
BPGM
Gene Name:
bisphosphoglycerate mutase
Gene Aliases:
-
Chromosome Location:
Chr.7: 13106850 - 13138358 on Build OryCun2.0
Species:
Rabbit
Species Specific ID (Flybase ID):
-
Assay ID Oc03823449_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BPGM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082269.2 NP_001075738.1 1 450 109
XM_008258120.2 XP_008256342.1 2 584 109
XM_008258121.2 XP_008256343.1 3 779 109
GenBank mRNA M15884.1 - 1 450 109

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

mutase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0