This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04900142_m1
See other TCAF2 GE Assays ›
Gene Symbol:
TCAF2
Gene Name:
TRPM8 channel associated factor 2
Gene Aliases:
-
Chromosome Location:
Chr.4: 115820763 - 115838098 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04900142_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TCAF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027968894.1 XP_027824695.1 8 - 9 2659 111
XM_027968895.1 XP_027824696.1 8 - 9 2656 111
XM_027968896.1 XP_027824697.1 8 - 9 3329 111
XM_027968898.1 XP_027824699.1 8 - 9 2638 111
XM_027968899.1 XP_027824700.1 8 - 9 3349 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0