This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04877415_g1
See other AKR1E2 GE Assays ›
Gene Symbol:
AKR1E2
Gene Name:
aldo-keto reductase family 1 member E2
Gene Aliases:
-
Chromosome Location:
Chr.13: 45431348 - 45457267 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04877415_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1E2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027976796.1 XP_027832597.1 6 - 7 683 52
XM_027976797.1 XP_027832598.1 6 - 7 634 52
XM_027976798.1 XP_027832599.1 6 - 7 673 52
XM_027976799.1 XP_027832600.1 5 - 6 530 52
XM_027976800.1 XP_027832601.1 6 - 7 683 52
XM_027976801.1 XP_027832602.1 6 - 7 683 52
XM_027976802.1 XP_027832603.1 6 - 7 767 52
XM_027976803.1 XP_027832604.1 6 - 7 767 52
XM_027976804.1 XP_027832605.1 6 - 7 767 52

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0