Assay Details
Gene Symbol: RPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963128.1 | XP_027818929.1 | 1 | 723 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: RPA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963128.1 | XP_027818929.1 | 1 | 723 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() |