Assay Details
Gene Symbol: SRRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017630.4 | XP_004017679.2 | 3 - 4 | 473 | 60 | |
XM_027956679.1 | XP_027812480.1 | 3 - 4 | 473 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
