This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04828991_m1
See other LMNTD1 GE Assays ›
Gene Symbol:
LMNTD1
Gene Name:
lamin tail domain containing 1
Gene Aliases:
IFLTD1
Chromosome Location:
Chr.3: 202802414 - 203316769 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04828991_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LMNTD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027967801.1 XP_027823602.1 15 - 16 2206 86
XM_027967802.1 XP_027823603.1 13 - 14 2051 86
XM_027967803.1 XP_027823604.1 13 - 14 1808 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0