Assay Details
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974231.1 | XP_027830032.1 | 4 - 5 | 298 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974231.1 | XP_027830032.1 | 4 - 5 | 298 | 59 |
Set Membership: |
![]() ![]() ![]() ![]() |