This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04676613_m1
See other CDKN3 GE Assays ›
Gene Symbol:
CDKN3
Gene Name:
cyclin dependent kinase inhibitor 3
Gene Aliases:
-
Chromosome Location:
Chr.7: 69090566 - 69108738 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04676613_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004010655.4 XP_004010704.2 7 - 8 624 74
XM_012181675.3 XP_012037065.2 7 - 8 621 74
XM_012181679.3 XP_012037069.2 7 - 8 489 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0