This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04657431_m1
See other APOBEC3Z1 GE Assays ›
Gene Symbol:
APOBEC3Z1
Gene Name:
apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like
Gene Aliases:
A3Z1
Chromosome Location:
Chr.3: 231362850 - 231366759 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa04657431_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APOBEC3Z1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001161379.1 NP_001154851.1 3 - 4 538 153
XM_027965728.1 XP_027821529.1 3 - 4 579 150
XM_027965729.1 XP_027821530.1 3 - 4 487 150
XM_027965730.1 XP_027821531.1 3 - 4 579 150
GenBank mRNA EU864541.1 - 3 - 4 538 153
KC513741.1 - 2 - 3 437 150
KC513742.1 - 2 - 3 440 153
KC522614.1 - 2 - 3 437 150

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Ovis aries

Panther Classification:

Molecular Function -

RNA processing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0