This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa04288537_m1
See other LOC114112156 GE Assays ›
Gene Symbol:
LOC114112156 DHX57
Gene Name:
putative ATP-dependent RNA helicase DHX57
DExH-box helicase 57
Gene Aliases:
-
-
Chromosome Location:
Chr.Unknown: 653 - 25515 on Build Oar_rambouillet_v1.0
Chromosome Location:
Chr.3: 90798600 - 90906576 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
-
Assay ID Oa04288537_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC114112156
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027963732.1 XP_027819533.1 3 - 4 1180 105
Gene Symbol: DHX57
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_027966915.1 XP_027822716.1 5 - 6 1472 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Ovis aries

Panther Classification:

Molecular Function -

RNA helicase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0