This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03253726_m1
See other TFAP2A GE Assays ›
Gene Symbol:
TFAP2A
Gene Name:
transcription factor AP-2 alpha
Gene Aliases:
AP2
Chromosome Location:
Chr.20: 49143529 - 49161855 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03253726_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TFAP2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001009745.1 NP_001009745.1 1 - 2 1653 70
XM_027958344.1 XP_027814145.1 2 - 3 674 70
XM_027958345.1 XP_027814146.1 2 - 3 1130 70
GenBank mRNA AF260568.1 - 2 - 3 533 70
AF260569.1 - 3 - 4 934 70
AF260570.1 - 1 - 2 1653 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0