This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03239618_m1
See other TENT2 GE Assays ›
Gene Symbol:
TENT2
Gene Name:
terminal nucleotidyltransferase 2
Gene Aliases:
PAPD4
Chromosome Location:
Chr.7: 11455882 - 11519804 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03239618_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TENT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004010211.3 XP_004010260.1 7 - 8 1397 77
XM_012180704.2 XP_012036094.1 8 - 9 1159 77
XM_012180705.2 XP_012036095.1 9 - 10 1299 77
XM_012180706.2 XP_012036096.1 8 - 9 899 77
XM_012180708.2 XP_012036098.1 8 - 9 1154 77
XM_012180710.3 XP_012036100.1 9 - 10 1039 77
XM_012180712.2 XP_012036102.1 8 - 9 1147 77
XM_027971490.1 XP_027827291.1 8 - 9 1159 77
XM_027971491.1 XP_027827292.1 4 - 5 1083 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0