This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03235610_m1
See other ERI3 GE Assays ›
Gene Symbol:
ERI3
Gene Name:
ERI1 exoribonuclease family member 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 19468088 - 19600542 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03235610_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERI3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004001901.3 XP_004001950.1 5 - 6 572 57
XM_012120995.3 XP_011976385.1 5 - 6 573 57
XM_012121058.3 XP_011976448.2 6 - 7 979 57
XM_012121260.3 XP_011976650.1 6 - 7 647 57
XM_012121548.2 XP_011976938.2 6 - 7 920 57
XM_027969053.1 XP_027824854.1 6 - 7 994 57
XM_027969058.1 XP_027824859.1 6 - 7 961 57
XM_027969075.1 XP_027824876.1 7 - 8 806 57
XM_027969090.1 XP_027824891.1 6 - 7 637 57
XM_027969091.1 XP_027824892.1 6 - 7 922 57
XM_027969092.1 XP_027824893.1 6 - 7 645 57
XM_027969094.1 XP_027824895.1 6 - 7 790 57

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries

Panther Classification:

Molecular Function -

exoribonuclease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0