Assay Details
Gene Symbol: EDNRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098228.2 | XP_014953714.2 | 4 - 5 | 1159 | 65 | |
GenBank mRNA | AF349439.1 | - | 1 - 2 | 35 | 65 | |
KY659245.1 | - | 4 - 5 | 1132 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
