This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Oa03210996_m1
See other MTFR1L GE Assays ›
Gene Symbol:
MTFR1L
Gene Name:
mitochondrial fission regulator 1 like
Gene Aliases:
FAM54B
Chromosome Location:
Chr.2: 256776790 - 256790632 on Build Oar_rambouillet_v1.0
Species:
Sheep
Species Specific ID (Flybase ID):
-
Assay ID Oa03210996_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MTFR1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_004005105.4 XP_004005154.1 3 - 4 341 71
XM_012151701.3 XP_012007091.1 3 - 4 369 71
XM_012151722.3 XP_012007112.1 3 - 4 411 71

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Ovis aries

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0