This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07306530_m1
See other U2AF1 GE Assays ›
Gene Symbol:
U2af1
Gene Name:
U2 small nuclear ribonucleoprotein auxiliary factor (U2AF) 1
Gene Aliases:
2010107D16Rik, 35kDa
Chromosome Location:
Chr.17: 31647081 - 31658764 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07306530_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: U2af1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024187.4 NP_077149.2 2 - 3 190 98
GenBank mRNA AK008332.1 - 2 - 3 152 98
AK012334.1 - 2 - 3 150 98
AK012849.1 - 2 - 3 150 98
AK088979.1 - 2 - 3 160 98
AK150872.1 - 2 - 3 190 98
BC002184.1 - 2 - 3 129 98
BC069888.1 - 2 - 3 132 98
BC115479.1 - 2 - 3 152 40100558 98
BC115480.1 - 2 - 3 152 40100559 98

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene U2af1

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
RNA splicing
nucleotide binding
nucleic acid binding
RNA binding
protein binding
metal ion binding
RS domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0