This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07304889_m1
See other CLDN10 GE Assays ›
Gene Symbol:
Cldn10
Gene Name:
claudin 10
Gene Aliases:
6720456I16Rik, Cldn10a, Cldn10b, D14Ertd728e
Chromosome Location:
Chr.14: 118787871 - 118874525 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07304889_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cldn10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001160097.1 NP_001153569.1 3 - 4 748 97
NM_001160098.1 NP_001153570.1 3 - 4 691 97
NM_001160099.1 NP_001153571.1 3 - 4 508 97
GenBank mRNA BC021770.1 - 3 - 4 701 5040100 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

tight junction

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
structural molecule activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0