This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07300166_g1
See other L2HGDH GE Assays ›
Gene Symbol:
L2hgdh
Gene Name:
L-2-hydroxyglutarate dehydrogenase
Gene Aliases:
BC016226
Chromosome Location:
Chr.12: 69690436 - 69724874 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07300166_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: L2hgdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145443.2 NP_663418.1 9 - 10 1276 93
GenBank mRNA AK145228.1 - 9 - 10 1275 93
AK152450.1 - 9 - 10 1253 93
AK168429.1 - 9 - 10 1224 93
BC016226.1 - 9 - 10 1220 4487011 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene L2hgdh

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

cellular protein metabolic process
oxidation-reduction process
oxidoreductase activity
2-hydroxyglutarate dehydrogenase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0