This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm07299009_s1
See other DDX28 GE Assays ›
Gene Symbol:
Ddx28
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 28
Gene Aliases:
2410004K13Rik, AI449652, Mddx28
Chromosome Location:
Chr.8: 106009616 - 106011486 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm07299009_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx28
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028038.3 NP_082314.2 1 1621 64
GenBank mRNA AK010396.1 - 1 1569 64
AK144945.1 - 1 2016 64
AK163480.1 - 1 1574 64
BC058568.1 - 1 1581 64
BC120556.1 - 1 1621 40129479 64
BC120582.1 - 1 1621 40129505 64

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx28

Gene Ontology Categories:

Function(s) Process(es)

biological_process
RNA secondary structure unwinding
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0